missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Cr, Alloy 690, N06690, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Nickel, Ni/Cr, Alloy 690, N06690, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Tungsten/Hafnium/Zinc/Lead/Tin/Boron/Niobium/Zirconium/Sulfur/Manganese/Copper/Tantalum/Oxygen/Phosphorus/Molybdenum/Cobalt/Vanadium/Silicon/Nitrogen/Carbon/Magnesium/Aluminium/Titanium/Iron/Chromium/Nickel/Rhenium |
| CAS | 7440-02-0 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Tungsten -60 ppm/Hafnium -20 ppm/Zinc -20 ppm/Lead -10 ppm/Tin -9 ppm/Boron 0.0005 %/Niobium 0.0011 %/Zirconium 0.0011 %/Sulfur 0.0013 %/Manganese 0.0017 %/Copper 0.002 %/Tantalum 0.002 %/Oxygen 0.0021 %/Phosphorus 0.0022 %/Molybdenum 0.0027 %/Cobalt 0.009 %/Vanadium 0.009 %/Silicon 0.011 %/Nitrogen 0.013 %/Carbon 0.014 %/Magnesium 0.015 %/Aluminium 0.32 %/Titanium 0.346 %/Iron 9.09 %/Chromium 29.5 %/Nickel 60.1 %/Rhenium (<40) ppm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction