missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Mo/Cr, Alloy 242, N10242, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Nickel, Ni/Mo/Cr, Alloy 242, N10242, 100g
Specifications
Specifications
| Chemical Name or Material | Phosphorus/Titanium/Carbon/Cobalt/Tungsten/Aluminium/Manganese/Iron/Chromium/Tin/Vanadium/Zirconium/Boron/Tantalum/Niobium/Copper/Molybdenum/Nickel |
| CAS | 7440-02-0 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Phosphorus 0.003 %/Titanium 0.01 %/Carbon 0.03 %/Cobalt 0.04 %/Tungsten 0.09 %/Aluminium 0.24 %/Manganese 0.33 %/Iron 1.09 %/Chromium 7.9 %/Tin (<0.01) %/Vanadium (<0.01) %/Zirconium (<0.02) %/Boron (0.005) %/Tantalum (0.01) %/Niobium (0.02) %/Copper (0.03) %/Molybdenum (25.4) %/Nickel (64.71) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction