Learn More
Ansell NITROTOUGH™ N250Y Medium Duty Oil-Repellant Gloves
Product Code. 16895310 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
11
Unit Size:
12 pairs
Product Code. 16895310
Brand: Ansell C261100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Nitrile coating all over the gloves provides a high level of oil repellance, while the elasticated knit wrist ensures a secure fit.
  • Nitrile coating provides oil repellence on the palm and back of the hand
  • Elasticated knitwrist ensures a secure fit
  • Comfortable and flexible
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Coating Material Nitrile
Color Yellow
Size 10
EN388 Standard 4111A
For Use With (Application) Assembly, General Maintenance, Metal Fabrication, and Quality Control
Glove Cuff Color Natural
Glove Cuff Style Knit
Liner Material 100% Cotton
Series N250Y