missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLK Antibody (SR2028), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22377-50ul
This item is not returnable.
View return policy
Description
NLK Monoclonal antibody specifically detects NLK in Human samples. It is validated for Western Blot, Flow Cytometry
Specifications
| NLK | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| SR2028 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
| DKFZp761G1211, EC 2.7.11, EC 2.7.11.24, FLJ21033, LAK1, nemo like kinase, nemo-like kinaseserine/threonine-protein kinase NLK, Protein LAK1 | |
| A synthesized peptide derived from human NLK (Uniprot #: Q9UBE8) | |
| 50 μg | |
| Protein Kinase, Wnt Signaling Pathway | |
| 51701 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction