missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLK Antibody (SR2028), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | NLK |
|---|---|
| Clone | SR2028 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18632884
|
Novus Biologicals
NBP3-22377-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18634214
|
Novus Biologicals
NBP3-22377-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NLK Monoclonal antibody specifically detects NLK in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
| NLK | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp761G1211, EC 2.7.11, EC 2.7.11.24, FLJ21033, LAK1, nemo like kinase, nemo-like kinaseserine/threonine-protein kinase NLK, Protein LAK1 | |
| A synthesized peptide derived from human NLK (Uniprot #: Q9UBE8) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2028 | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase, Wnt Signaling Pathway | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 51701 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title