Learn More
Nociceptin Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93943-0.02ml
This item is not returnable.
View return policy
Description
Nociceptin Polyclonal antibody specifically detects Nociceptin in Human samples. It is validated for Western Blot
Specifications
| Nociceptin | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| nociceptin, nocistatin, OFQ, P, PPNOC, prepronociceptin, propronociceptin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-176 of human PNOC (NP_006219.1). SCQRDCLTCQEKLHPALDSFDLEVCILECEEKVFPSPLWTPCTKVMARSSWQLSPAAPEHVAAALYQPRASEMQHLRRMPRVRSLFQEQEEPEPGMEEAGEMEQKQLQKRFGGFTGARKSARKLANQKRFSEFMRQYLVLSMQSSQRRRTLHQNGNV | |
| 0.02 mL | |
| Neuroscience, Signal Transduction | |
| 5368 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |