missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nociceptin Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 470.00
Specifications
| Antigen | Nociceptin |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18638211
|
Novus Biologicals
NBP2-93943-0.02ml |
0.02 mL |
€ 188.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18685551
|
Novus Biologicals
NBP2-93943-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nociceptin Polyclonal antibody specifically detects Nociceptin in Human samples. It is validated for Western BlotSpécification
| Nociceptin | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 5368 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| nociceptin, nocistatin, OFQ, P, PPNOC, prepronociceptin, propronociceptin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-176 of human PNOC (NP_006219.1). SCQRDCLTCQEKLHPALDSFDLEVCILECEEKVFPSPLWTPCTKVMARSSWQLSPAAPEHVAAALYQPRASEMQHLRRMPRVRSLFQEQEEPEPGMEEAGEMEQKQLQKRFGGFTGARKSARKLANQKRFSEFMRQYLVLSMQSSQRRRTLHQNGNV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit