missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nogo B receptor Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-24950
This item is not returnable.
View return policy
Description
Nogo B receptor Polyclonal specifically detects Nogo B receptor in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Nogo B receptor | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| NUS1 | |
| A synthetic peptide corresponding to amino acids 64-74 (RNRRHHRHPRG) of human Nogo-B receptor (NgBR) was used as immunogen, GenBank no sp Q96E22.1 NGBR_HUMAN (Miao et al, 2006). This immunogen sequence is within the 93 amino acid putative ectodomain of the NgBR. | |
| 0.1 mg | |
| Lipid and Metabolism | |
| 116150 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1-3 ug/ml | |
| C6orf68MGC117249, chromosome 6 open reading frame 68, MGC:7199, MGC7199, NgBR, nogo-B receptor, Nuclear undecaprenyl pyrophosphate synthase 1 homolog, nuclear undecaprenyl pyrophosphate synthase 1 homolog (S. cerevisiae) | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction