missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nogo B receptor Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 213.00 - € 453.00
Specifications
| Antigen | Nogo B receptor |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1-3 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18103204
|
Novus Biologicals
NBP2-24950SS |
0.025 mg |
€ 213.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18757493
|
Novus Biologicals
NBP2-24950 |
0.1 mg |
€ 453.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nogo B receptor Polyclonal specifically detects Nogo B receptor in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| Nogo B receptor | |
| Western Blot 1-3 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| C6orf68MGC117249, chromosome 6 open reading frame 68, MGC:7199, MGC7199, NgBR, nogo-B receptor, Nuclear undecaprenyl pyrophosphate synthase 1 homolog, nuclear undecaprenyl pyrophosphate synthase 1 homolog (S. cerevisiae) | |
| NUS1 | |
| IgG | |
| Protein G purified |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 116150 | |
| A synthetic peptide corresponding to amino acids 64-74 (RNRRHHRHPRG) of human Nogo-B receptor (NgBR) was used as immunogen, GenBank no sp Q96E22.1 NGBR_HUMAN (Miao et al, 2006). This immunogen sequence is within the 93 amino acid putative ectodomain of the NgBR. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title