missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NONO Antibody (8C11), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26110-50ul
This item is not returnable.
View return policy
Description
NONO Monoclonal antibody specifically detects NONO in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry
Specifications
| NONO | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry | |
| 8C11 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| NMT5552 kDa subunit, non-POU domain containing, octamer-binding, NRB54non-POU-domain-containing, octamer-binding, p54(nrb) | |
| A synthesized peptide derived from Human NONO [UniProt Q15233] | |
| 50 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 4841 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction