Learn More
Honeywell™ North™ NorthFlex Cold Grip™ PVC-Coated Polyamide Insulated Gloves
Product Code. 15545006
Click to view available options
Size:
Large
X-Large
Unit Size:
12 pairs
Product Code. 15545006
Brand: Honeywell™ North™ NF11HD/10XL
Only null left
Add to basket
Add to basket
Cold Grip™ Gloves provide workers with comfort and enhanced grip capabilities, while also offering protection from temperatures as low as -26°C (-15°F).
  • Heavyweight insulated gloves without sacrificing comfort and dexterity
  • Polyamide yarn reduces linting for the sensitive applications
  • Black PVC coating covers the palm and fingers for enhanced grip capabilities
  • Hydropellent Technology repels liquids to provide a dry, firm grip
  • Ideal for handling damp, oily, or greasy parts in cold conditions
  • Recommended for temperatures down to -26°C (-15°F)
  • Launderable
TRUSTED_SUSTAINABILITY
Specifications
Coating Material PVC
Color Black
Exterior Finish Fully Textured
Hand Specific
Material Polyamide
Size X-Large
For Use With (Application) Construction, Logistics, and Maintenance
Glove Cuff Color Blue
Glove Cuff Style Knit
Liner Material Synthetic Insulation
Series NF11HD
Temperature (English) Min -15°F
Temperature (Metric) Min. -26°C
Yarn Gauge 7