Learn More
Notch-2 Antibody (B6) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-62559
 View more versions of this product
Product Code. 18661908
  • € 602.00 / 0.20mg
Description
Notch-2 Monoclonal specifically detects Notch-2 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Notch-2
Monoclonal
Unconjugated
PBS
NOTCH2
NRR of mouse Notch 2
4853
Human, Mouse
Purified
ELISA, Immunohistochemistry, Immunofluorescence
B6
ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunofluorescence
AGS2, hN2, neurogenic locus notch homolog protein 2, Notch (Drosophila) homolog 2, notch 2Notch homolog 2 (Drosophila), Notch homolog 2
Rabbit
0.2 mg
An engineered ScFv fragment antibody that binds to both mouse mNRR2 and human hNRR2, but not NRR1.
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
IgG κ