missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notch-2 Antibody (B6) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 282.00 - € 602.00
Specifications
| Antigen | Notch-2 |
|---|---|
| Clone | B6 |
| Dilution | ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunofluorescence |
| Applications | ELISA, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18023386
|
Novus Biologicals
NBP2-62559-0.025MG |
0.025 mg |
€ 282.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18661908
|
Novus Biologicals
NBP2-62559 |
0.2 mg |
€ 602.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Notch-2 Monoclonal specifically detects Notch-2 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence.Specifications
| Notch-2 | |
| ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunofluorescence | |
| Monoclonal | |
| Purified | |
| Human, Mouse | |
| AGS2, hN2, neurogenic locus notch homolog protein 2, Notch (Drosophila) homolog 2, notch 2Notch homolog 2 (Drosophila), Notch homolog 2 | |
| NOTCH2 | |
| IgG κ | |
| An engineered ScFv fragment antibody that binds to both mouse mNRR2 and human hNRR2, but not NRR1. |
| B6 | |
| ELISA, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| PBS | |
| 4853 | |
| NRR of mouse Notch 2 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title