missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notch-3 Antibody (2E4D11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-52521-0.025mg
This item is not returnable.
View return policy
Description
Notch-3 Monoclonal specifically detects Notch-3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
Specifications
| Notch-3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence, CyTOF-ready | |
| CADASIL, CASILneurogenic locus notch homolog protein 3, Notch (Drosophila) homolog 3, notch 3Notch homolog 3 (Drosophila), Notch homolog 3 | |
| Mouse | |
| 243.6 kDa | |
| 0.025 mg | |
| Cancer | |
| 4854 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF | |
| 2E4D11 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| NOTCH3 | |
| Purified recombinant fragment of human Notch3 (AA: 47-156) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction