missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notch-3 Antibody (2E4D11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
€ 230.00 - € 578.00
Specifications
| Antigen | Notch-3 |
|---|---|
| Clone | 2E4D11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence, CyTOF-ready |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18281021
|
Novus Biologicals
NBP2-52521 |
0.1 mg |
€ 578.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663029
|
Novus Biologicals
NBP2-52521-0.025mg |
0.025 mg |
€ 230.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Notch-3 Monoclonal specifically detects Notch-3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
| Notch-3 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| PBS with 0.05% Sodium Azide | |
| 4854 | |
| Purified recombinant fragment of human Notch3 (AA: 47-156) expressed in E. Coli. | |
| Primary | |
| 243.6 kDa |
| 2E4D11 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| CADASIL, CASILneurogenic locus notch homolog protein 3, Notch (Drosophila) homolog 3, notch 3Notch homolog 3 (Drosophila), Notch homolog 3 | |
| NOTCH3 | |
| IgG2b | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title