Learn More
Fisherbrand™ NOVOsil™ Translucent Silicone Tubing
Product Code. p-8006498 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Inner:
3 mm
4 mm
5 mm
8 mm
10 mm
Unit Size:
Each
Product Code. 11788705
Only null left
Add to basket
Add to basket
Providing good elasticity and suitable for increased mechanical loads in the laboratory. Fisherbrand™ NOVOsil™ Translucent Silicone Tubing is neutral, non-toxic and delivered free of talcum.
  • Autoclavable
  • Flexible and soft
  • Suitable for hose pumps
  • Resistant to mold, water, radiation and light effects
  • Excellent compatibility with sterile solutions
  • Applications include pharmaceutical and biotechnology
  • Not recommended for concentrated solvents, oils, diluted acids and alkalis
Specifications
Certifications/Compliance FDA, BFR and WRAS approved
Autoclavable Autoclavable
Diameter (Metric) Inner 3 mm
Material Silicone
Diameter (Metric) Outer 7 mm
Overall Length (Metric) 25 m
For Use With (Application) Ideal for pharmaceutical, biotechnology, sterile fluids transfers
Min. Operating Temperature (Metric) -40°C
Color Translucent
Flexibility Flexible
Maximum Pressure 1.5 bar
Max. Operating Temperature (Metric) 250°C