missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPY5R Antibody (SR1903), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22491-100ul
This item is not returnable.
View return policy
Description
NPY5R Monoclonal antibody specifically detects NPY5R in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
Specifications
| NPY5R | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| SR1903 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| neuropeptide Y receptor type 5, neuropeptide Y receptor Y5, NPYR5NPY5-R, NPY-Y5 receptor, NPYY5-R, Y5 receptor | |
| A synthesized peptide derived from human NPY5R (Uniprot #: Q15761) | |
| 100 μg | |
| GPCR, Neuroscience, Neurotransmission | |
| 4889 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction