missing translation for 'onlineSavingsMsg'
Learn More

NPY5R Antibody (SR1903), Novus Biologicals™

Rabbit Monoclonal Antibody

€ 349.00 - € 472.00

Specifications

Antigen NPY5R
Clone SR1903
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18652394
View Documents
Novus Biologicals
NBP3-22491-100ul
100 μg
€ 472.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18627604
View Documents
Novus Biologicals
NBP3-22491-50ul
50 μg
€ 349.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NPY5R Monoclonal antibody specifically detects NPY5R in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
Specifications

Specifications

NPY5R
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200
Monoclonal
Purified
RUO
Human
neuropeptide Y receptor type 5, neuropeptide Y receptor Y5, NPYR5NPY5-R, NPY-Y5 receptor, NPYY5-R, Y5 receptor
A synthesized peptide derived from human NPY5R (Uniprot #: Q15761)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR1903
Western Blot, Flow Cytometry, Immunofluorescence
Unconjugated
Rabbit
GPCR, Neuroscience, Neurotransmission
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
4889
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.