missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NQO2 Monoclonal Antibody (OTI5F6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504782S
This item is not returnable.
View return policy
Description
NQO2 Monoclonal antibody specifically detects NQO2 in Canine, Human, Mouse samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| NQO2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P16083, Q9JI75 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 18105, 4835, 606932 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Western Blot | |
| OTI5F6 | |
| Unconjugated | |
| NQO2 | |
| DHQV; DIA6; EC 1.10.99.2; MGC94180; NAD(P)H dehydrogenase, quinone 2; NAD(P)H menadione oxidoreductase 2, dioxin inducible; NAD(P)H menadione oxidoreductase-1, dioxin-inducible-2; NAD(P)H quinone dehydrogenase 2; NMO2; NMOR2; NQO 2; Nqo2; NRH dehydrogenase; NRH dehydrogenase [quinone] 2; NRH: quinone oxidoreductase; NRH:quinone oxidoreductase 2; N-ribosyldihydronicotinamide:quinone reductase 2; OTTHUMP00000015948; OTTHUMP00000015949; OTTHUMP00000015953; Ox 2; Ox2; QR2; Quinone; Quinone reductase 2; quinone ribosyldihydronicotinamide dehydrogenase; ribosyldihydronicotinamide; ribosyldihydronicotinamide dehydrogenase; ribosyldihydronicotinamide dehydrogenase [quinone]; ribosyldihydronicotinamide dehydrogenase quinone | |
| Full length human recombinant protein of human NQO2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human, Mouse | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction