Learn More
NQO2 Monoclonal Antibody (OTI5F6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA504782
Encompass_Preferred
 View more versions of this product
Product Code. 17512412
  • € 540.87 / 100µL
Description
NQO2 (EC 1.10.99.2) is a flavoprotein that catalyzes the 2-electron reduction of various quinones, redox dyes, and the vitamin K menadione. NQO2 predomitly uses dihydronicotinamide riboside (NRH) as the electron donor.
Specifications
Specifications
NQO2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P16083
NQO2
Full length human recombit protein of human NQO2 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Western Blot
OTI5F6
Unconjugated
NQO2
DHQV, DIA6, NMOR2, QR2
Mouse
Affinity Chromatography
RUO
4835
-20° C, Avoid Freeze/Thaw Cycles