missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NSMAF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30929-25ug
This item is not returnable.
View return policy
Description
NSMAF Polyclonal antibody specifically detects NSMAF in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| NSMAF | |
| Polyclonal | |
| Western Blot : 1:400, Immunohistochemistry : 10-15 μg/mL | |
| Factor associated with neutral sphingomyelinase activation, FANFactor associated with N-SMase activation, neutral sphingomyelinase (N-SMase) activation associated factor, protein FAN | |
| Synthetic peptide derived from human NSMAF protein. | |
| 25 μg | |
| Cell Biology, Immunology, Innate Immunity, Signal Transduction | |
| 8439 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction