missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NSMAF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 213.00 - € 508.00
Specifications
| Antigen | NSMAF |
|---|---|
| Dilution | Western Blot : 1:400, Immunohistochemistry : 10-15 μg/mL |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
30067060
|
Novus Biologicals™
NBP3-30929-100ug |
100 μg |
€ 508.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066889
|
Novus Biologicals™
NBP3-30929-25ug |
25 μg |
€ 213.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
NSMAF Polyclonal antibody specifically detects NSMAF in Human samples. It is validated for Immunohistochemistry,Western BlotSpezifikation
| NSMAF | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Immunology, Innate Immunity, Signal Transduction | |
| Phosphate buffered saline | |
| 8439 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot : 1:400, Immunohistochemistry : 10-15 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Factor associated with neutral sphingomyelinase activation, FANFactor associated with N-SMase activation, neutral sphingomyelinase (N-SMase) activation associated factor, protein FAN | |
| Synthetic peptide derived from human NSMAF protein. | |
| Antigen Affinity-purified |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts