missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleolin Antibody (NCL/7014R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-11037-20ug
This item is not returnable.
View return policy
Description
Nucleolin Monoclonal antibody specifically detects Nucleolin in Human, Bovine (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Nucleolin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 μg/ml, Flow Cytometry 1-2 μg/million cells, Immunohistochemistry 1-2ug/ml, Immunocytochemistry/ Immunofluorescence 1-2 μg/ml, Immunohistochemistry-Paraffin 1-2 μg/ml | |
| P19338 | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Bovine (Negative), Mouse (Negative), Rat (Negative) | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| NCL/7014R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| C23, FLJ45706, FLJ59041, nucleolin, Protein C23 | |
| Recombinant full-length human Nucleolin protein (Uniprot: P19338) | |
| 20 μg | |
| Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers | |
| 4691 | |
| Store at 4°C. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction