missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleolin Antibody (NCL/7014R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 318.00 - € 559.00
Specifications
| Antigen | Nucleolin |
|---|---|
| Clone | NCL/7014R |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 μg/ml, Flow Cytometry 1-2 μg/million cells, Immunohistochemistry 1-2ug/ml, Immunocytochemistry/ Immunofluorescence 1-2 μg/ml, Immunohistochemistry-Paraffin 1-2 μg/ml |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18657347
|
Novus Biologicals
NBP3-11037-20ug |
20 μg |
€ 318.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18606608
|
Novus Biologicals
NBP3-11037-100ug |
100 μg |
€ 559.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nucleolin Monoclonal antibody specifically detects Nucleolin in Human, Bovine (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Nucleolin | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| C23, FLJ45706, FLJ59041, nucleolin, Protein C23 | |
| Recombinant full-length human Nucleolin protein (Uniprot: P19338) | |
| Primary | |
| Store at 4°C. |
| NCL/7014R | |
| Western Blot 1-2 μg/ml, Flow Cytometry 1-2 μg/million cells, Immunohistochemistry 1-2ug/ml, Immunocytochemistry/ Immunofluorescence 1-2 μg/ml, Immunohistochemistry-Paraffin 1-2 μg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Bovine (Negative), Mouse (Negative), Rat (Negative) | |
| P19338 | |
| 4691 | |
| IgG κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title