missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleolin, p Thr84 Antibody (SR1020), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21606-50ul
This item is not returnable.
View return policy
Description
Nucleolin Monoclonal antibody specifically detects Nucleolin in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Nucleolin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| Western Blot, Immunohistochemistry | |
| SR1020 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| C23, FLJ45706, FLJ59041, nucleolin, Protein C23 | |
| A synthesized peptide derived from human phospho-Nucleolin [Thr84] (Uniprot number: P19338) | |
| 50 μL | |
| Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific, Stem Cell Markers | |
| 4691 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction