missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleolin, p Thr84 Antibody (SR1020), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 373.00 - € 498.00
Specifications
| Antigen | Nucleolin |
|---|---|
| Clone | SR1020 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18603825
|
Novus Biologicals
NBP3-21606-100ul |
100 μg |
€ 498.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18616864
|
Novus Biologicals
NBP3-21606-50ul |
50 μL |
€ 373.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nucleolin Monoclonal antibody specifically detects Nucleolin in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| Nucleolin | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 4691 | |
| Affinity purified |
| SR1020 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific, Stem Cell Markers | |
| C23, FLJ45706, FLJ59041, nucleolin, Protein C23 | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title