missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleostemin Antibody (2C8D5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37410-0.025ml
This item is not returnable.
View return policy
Description
Nucleostemin Monoclonal specifically detects Nucleostemin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| Nucleostemin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| C77032, E2IG3NNP47, estradiol-induced nucleotide binding protein, guanine nucleotide binding protein-like 3 (nucleolar), guanine nucleotide-binding protein-like 3, MGC800, Novel nucleolar protein 47, NSE2-induced gene 3 protein, Nucleolar GTP-binding protein 3, nucleostemin | |
| Mouse | |
| 62 kDa | |
| 0.025 mL | |
| Apoptosis, Cellular Markers, Chromatin Modifiers, Core ESC Like Genes, Embryonic Stem Cell Markers, Epigenetics, Histones and Modified Histones, Metabotropic Glutamate Receptors, microRNA, mTOR Pathway, Neurofilaments, Stem Cell Markers, Stem Cell Signaling Pathway | |
| 26354 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 2C8D5 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| GNL3 | |
| Purified recombinant fragment of human GNL3 (AA: 1-226) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction