missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleostemin Antibody (2C8D5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 203.00 - € 533.00
Specifications
| Antigen | Nucleostemin |
|---|---|
| Clone | 2C8D5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18679885
|
Novus Biologicals
NBP2-37410-0.025ml |
0.025 mL |
€ 203.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18135059
|
Novus Biologicals
NBP2-37410 |
0.1 mL |
€ 533.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nucleostemin Monoclonal specifically detects Nucleostemin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| Nucleostemin | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| C77032, E2IG3NNP47, estradiol-induced nucleotide binding protein, guanine nucleotide binding protein-like 3 (nucleolar), guanine nucleotide-binding protein-like 3, MGC800, Novel nucleolar protein 47, NSE2-induced gene 3 protein, Nucleolar GTP-binding protein 3, nucleostemin | |
| GNL3 | |
| IgG1 | |
| Affinity Purified |
| 2C8D5 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Apoptosis, Cellular Markers, Chromatin Modifiers, Core ESC Like Genes, Embryonic Stem Cell Markers, Epigenetics, Histones and Modified Histones, Metabotropic Glutamate Receptors, microRNA, mTOR Pathway, Neurofilaments, Stem Cell Markers, Stem Cell Signaling Pathway | |
| PBS with 0.05% Sodium Azide | |
| 26354 | |
| Purified recombinant fragment of human GNL3 (AA: 1-226) expressed in E. Coli. | |
| Primary | |
| 62 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title