missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT19 Antibody (SR1715), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | NUDT19 |
|---|---|
| Clone | SR1715 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18602675
|
Novus Biologicals
NBP3-22576-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18682575
|
Novus Biologicals
NBP3-22576-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NUDT19 Monoclonal antibody specifically detects NUDT19 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| NUDT19 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| 390916 | |
| IgG | |
| Affinity purified |
| SR1715 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| EC 3.6.1, Nucleoside Diphosphate-Linked Moiety X Motif 19, Mitochondrial, Nudix (Nucleoside Diphosphate Linked Moiety X)-Type Motif 19, Nudix Motif 19, RP2 | |
| A synthesized peptide derived from human NUDT19 (Uniprot #: A8MXV4) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title