missing translation for 'onlineSavingsMsg'
Learn More

NUDT19 Antibody (SR1715), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-22576-50ul

 View more versions of this product

Product Code. 18682575

  • € 338.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NUDT19 Monoclonal antibody specifically detects NUDT19 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NUDT19
Monoclonal
Unconjugated
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol
Rabbit
Affinity purified
RUO
390916
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunofluorescence
SR1715
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200
EC 3.6.1, Nucleoside Diphosphate-Linked Moiety X Motif 19, Mitochondrial, Nudix (Nucleoside Diphosphate Linked Moiety X)-Type Motif 19, Nudix Motif 19, RP2
A synthesized peptide derived from human NUDT19 (Uniprot #: A8MXV4)
50 μg
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.