missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT22 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98534-50ul
This item is not returnable.
View return policy
Description
NUDT22 Polyclonal antibody specifically detects NUDT22 in Human samples. It is validated for Western Blot
Specifications
| NUDT22 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| MGC13045, nucleoside diphosphate-linked moiety X motif 22, nudix (nucleoside diphosphate linked moiety X)-type motif 22, Nudix motif 22 | |
| Produced in rabbits immunized with E. coli-derived Human NUDT22 fragment. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 84304 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction