missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NULP1 Mouse anti-Human, Clone: PCRP-TCF25-1A11, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07359-100ug
This item is not returnable.
View return policy
Description
NULP1 Monoclonal specifically detects NULP1 in Human samples. It is validated for Western Blot, Flow Cytometry, Protein Array.
Specifications
| NULP1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Protein Array | |
| hKIAA1049, KIAA1049Hulp1, Nuclear localized protein 1, Nulp1, NULP1PRO2620, TCF-25, transcription factor 25, transcription factor 25 (basic helix-loop-helix) | |
| Recombinant full-length human NULP1 protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Peptide Array | |
| PCRP-TCF25-1A11 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 22980 | |
| Store at 4°C. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction