missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NULP1 Mouse anti-Human, Clone: PCRP-TCF25-1A11, Novus Biologicals™
Mouse Monoclonal Antibody
€ 484.00
Specifications
| Antigen | NULP1 |
|---|---|
| Clone | PCRP-TCF25-1A11 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Protein Array |
| Applications | Western Blot, Flow Cytometry, Peptide Array |
Description
NULP1 Monoclonal specifically detects NULP1 in Human samples. It is validated for Western Blot, Flow Cytometry, Protein Array.Specifications
| NULP1 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| hKIAA1049, KIAA1049Hulp1, Nuclear localized protein 1, Nulp1, NULP1PRO2620, TCF-25, transcription factor 25, transcription factor 25 (basic helix-loop-helix) | |
| Recombinant full-length human NULP1 protein | |
| Primary | |
| Store at 4°C. |
| PCRP-TCF25-1A11 | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 22980 | |
| IgG2a | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title