missing translation for 'onlineSavingsMsg'
Learn More

NuMA Antibody (12A11), Novus Biologicals™

Rabbit Monoclonal Antibody

€ 343.00 - € 533.00

Specifications

Antigen NuMA
Clone 12A11
Dilution Flow Cytometry 1:50-1:200, ELISA
Applications Flow Cytometry, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30027672
View Documents
Novus Biologicals
NBP3-26451-50ul
50 μL
€ 343.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30034487
View Documents
Novus Biologicals
NBP3-26451-100ul
100 μL
€ 533.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NuMA Monoclonal antibody specifically detects NuMA in Human samples. It is validated for Flow Cytometry, ELISA
Specifications

Specifications

NuMA
Flow Cytometry 1:50-1:200, ELISA
Monoclonal
Purified
RUO
Human
centrophilin stabilizes mitotic spindle in mitotic cells, nuclear mitotic apparatus protein 1, NUMA, NuMA protein, SP-H antigen, structural nuclear protein
A synthesized peptide derived from Human NuMA [UniProt Q14980]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
12A11
Flow Cytometry, ELISA
Unconjugated
Rabbit
Breast Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Mitotic Regulators
PBS, pH 7.4, 150mM NaCl and 50% glycerol
4926
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.