Learn More
Thermo Scientific™ Nunc™ General Long Term Storage Tubes
Product Code. p-4526577
Click to view available options
Volume (Metric):
1.8 mL
4.5 mL
Unit Size:
Pack of 1200
Pack of 1800
Product Code. 10726662
Brand: Thermo Scientific™ 347783
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Contain samples in cost-efficient round-bottom, externally-threaded Thermo Scientific™ Nunc™ General Long Term Storage Tubes.
Contiain samples in cost-efficient round-bottom, externally-threaded Thermo Scientific™ Nunc™ General Long Term Storage Tubes.
Starfoot vial fits into grooves of the CryoTube Rack (Cat. No. 376589), which allows one hand operation.
  • Caps and tubes packaged separately to save handling time
  • Tubes are nonsterile
  • Economical choice for less demanding applications
TRUSTED_SUSTAINABILITY
Specifications
Description Nunc Stor-It Tubes, external thread, round bottom
Color White
Diameter (Metric) Outer 48 mm
Sterility Non-sterile
Volume (Metric) 1.8 mL
No. per Case 1800
No. per Pack 450
Style Starfoot, round-bottom
Volume (Metric) Total 1.8 mL
Height (Metric) 12.5 mm
Quantity 1800/Cs., 450/Pk.
Type Storage Tube