Learn More
Invitrogen™ NUP155 Recombinant Rabbit Monoclonal Antibody (23GB2010), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553197
 View more versions of this product
Product Code. 30088130
  • € 217.00 / 20µL
Description
Nucleoporins are the main components of the nuclear pore complex (NPC) of eukaryotic cells. They are involved in the bidirectional trafficking of molecules, especially mRNAs and proteins, between the nucleus and the cytoplasm. The protein does not contain the typical FG repeat sequences found in most vertebrate nucleoporins.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NUP155
Recombinant Monoclonal
1.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O75694, P37199, Q99P88
NUP155
A synthesized peptide derived from human NUP155 (901-1050AA).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB2010
Unconjugated
NUP155
155 kDa nucleoporin; ATFB15; D930027M19Rik; KIAA0791; mKIAA0791; N155; Nuclear pore complex protein Nup155; nucleoporin 155; nucleoporin 155kD; nucleoporin 155kDa; nucleoporin Nup155; NUP155; P140
Rabbit
Affinity chromatography
RUO
117021, 170762, 9631
-20°C