missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUP155 Recombinant Rabbit Monoclonal Antibody (23GB2010), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 217.00 - € 590.00
Specifications
| Antigen | NUP155 |
|---|---|
| Clone | 23GB2010 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088117
![]() |
Invitrogen™
MA553196 |
100 μL |
€ 590.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088130
|
Invitrogen™
MA553197 |
20 μL |
€ 217.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NUP155 Recombinant Monoclonal antibody specifically detects NUP155 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| NUP155 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O75694, P37199, Q99P88 | |
| 117021, 170762, 9631 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2010 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 155 kDa nucleoporin; ATFB15; D930027M19Rik; KIAA0791; mKIAA0791; N155; Nuclear pore complex protein Nup155; nucleoporin 155; nucleoporin 155kD; nucleoporin 155kDa; nucleoporin Nup155; NUP155; P140 | |
| A synthesized peptide derived from human NUP155 (901-1050AA). | |
| Primary | |
| -20°C | |
| NUP155 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title