missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUP155 Recombinant Rabbit Monoclonal Antibody (23GB2010), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553196
This item is not returnable.
View return policy
Description
Nucleoporins are the main components of the nuclear pore complex (NPC) of eukaryotic cells. They are involved in the bidirectional trafficking of molecules, especially mRNAs and proteins, between the nucleus and the cytoplasm. The protein does not contain the typical FG repeat sequences found in most vertebrate nucleoporins.
Specifications
| NUP155 | |
| Recombinant Monoclonal | |
| 1.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75694, P37199, Q99P88 | |
| NUP155 | |
| A synthesized peptide derived from human NUP155 (901-1050AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2010 | |
| Unconjugated | |
| NUP155 | |
| 155 kDa nucleoporin; ATFB15; D930027M19Rik; KIAA0791; mKIAA0791; N155; Nuclear pore complex protein Nup155; nucleoporin 155; nucleoporin 155kD; nucleoporin 155kDa; nucleoporin Nup155; NUP155; P140 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 117021, 170762, 9631 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction