Learn More
NUP62CL Antibody (S08-1A5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19818-100ul
This item is not returnable.
View return policy
Description
NUP62CL Monoclonal antibody specifically detects NUP62CL in Human samples. It is validated for Western Blot
Specifications
| NUP62CL | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| S08-1A5 | |
| Unconjugated | |
| FLJ20130, nucleoporin 62kDa C-terminal like, nucleoporin-62 C-terminal-like protein, NUP62L | |
| A synthetic peptide of human NUP62CL (Uniprot # Q9H1M0) | |
| 100 μg | |
| DNA replication Transcription Translation and Splicing | |
| 54830 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |