missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP62CL Antibody (S08-1A5), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 421.00 - € 645.00
Specifications
| Antigen | NUP62CL |
|---|---|
| Clone | S08-1A5 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18637433
|
Novus Biologicals
NBP3-19818-50ul |
50 μg |
€ 421.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18608573
|
Novus Biologicals
NBP3-19818-100ul |
100 μg |
€ 645.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NUP62CL Monoclonal antibody specifically detects NUP62CL in Human samples. It is validated for Western BlotSpecifications
| NUP62CL | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ20130, nucleoporin 62kDa C-terminal like, nucleoporin-62 C-terminal-like protein, NUP62L | |
| A synthetic peptide of human NUP62CL (Uniprot # Q9H1M0) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S08-1A5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 54830 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title