missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP98 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33379-100ul
This item is not returnable.
View return policy
Description
NUP98 Monoclonal antibody specifically detects NUP98 in Human,Monkey samples. It is validated for ELISA,Western Blot
Specifications
| NUP98 | |
| Monoclonal | |
| Western Blot 1:2000 - 1:7000, ELISA Recommended starting concentration is 1 μg/mL | |
| ADAR2, ADIR2, GLFG-repeat containing nucleoporin, nuclear pore complex protein Nup98-Nup96, nucleoporin 98kD, nucleoporin 98kDa, NUP196, NUP96, Nup98-Nup96 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 712-863 of human NUP98 (NP_001352055.1).,, Sequence:, DESLQDDREEIENNSYHMHPAGIILTKVGYYTIPSMDDLAKITNEKGECIVSDFTIGRKGYGSIYFEGDVNLTNLNLDDIVHIRRKEVVVYLDDNQKPPVGEGLNRKAEVTLDGVWPTDKTSRCLIKSPDRLADINYEGRLEAVSRKQGAQF | |
| 100 μL | |
| Cancer, Mitotic Regulators | |
| 4928 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction