missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP98 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 213.00 - € 507.00
Specifications
| Antigen | NUP98 |
|---|---|
| Dilution | Western Blot 1:2000 - 1:7000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227143
|
Novus Biologicals
NBP3-33379-20ul |
20 μL |
€ 213.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229846
|
Novus Biologicals
NBP3-33379-100ul |
100 μL |
€ 507.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NUP98 Monoclonal antibody specifically detects NUP98 in Human,Monkey samples. It is validated for ELISA,Western BlotSpecifications
| NUP98 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Mitotic Regulators | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 4928 | |
| IgG | |
| Affinity purified |
| Western Blot 1:2000 - 1:7000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Monkey | |
| ADAR2, ADIR2, GLFG-repeat containing nucleoporin, nuclear pore complex protein Nup98-Nup96, nucleoporin 98kD, nucleoporin 98kDa, NUP196, NUP96, Nup98-Nup96 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 712-863 of human NUP98 (NP_001352055.1).,, Sequence:, DESLQDDREEIENNSYHMHPAGIILTKVGYYTIPSMDDLAKITNEKGECIVSDFTIGRKGYGSIYFEGDVNLTNLNLDDIVHIRRKEVVVYLDDNQKPPVGEGLNRKAEVTLDGVWPTDKTSRCLIKSPDRLADINYEGRLEAVSRKQGAQF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title