Learn More
Trajan Scientific and Medical™ Nuts for Fittings
Gives low dead volume connections for optimum chromatography.
Brand:  Trajan Scientific and Medical™ 1034050
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15249113
  • € 111.00 / Pack of 5
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
  • Low dead volume
  • High temperature applications
  • Leak free connections
Specifications
Specifications
3.17 mm Hole (Extended)
Stainless Steel
Nuts for Fittings 1/8 in. Hole (Extended)
5/Pk.