Learn More
Trajan Scientific and Medical™ Nuts for Fittings
Gives low dead volume connections for optimum chromatography.
€ 47.50 - € 111.00
Product Code | Brand | Size (Metric) | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size (Metric) | Material | Price | Quantity & Availability | ||||
11987957
|
Trajan Scientific and Medical™
103406 |
1.59 mm x 1.2 mm Hole (Extended) | Nickel-plated Brass |
€ 47.50
Pack of 5 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
15249113
|
Trajan Scientific and Medical™
1034050 |
3.17 mm Hole (Extended) | Stainless Steel |
€ 111.00
Pack of 5 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
Description
- Low dead volume
- High temperature applications
- Leak free connections
Specifications
5/Pk. |