Learn More
Occludin Rabbit, anti-Human, CoraLite Plus 488, Clone: 8B2, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand:  Proteintech CL488-80545-20UL
 View more versions of this product
Product Code. 18077275
  • € 145.00 / 20µL
Description
The Occludin antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Occludin. This antibody recognizes human, mouse, pig antigen. The Occludin antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
Occludin
8B2
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Pig
150 kDa
IgG
Immunofluorescence
1000 μg/mL
occludin, OCLN
20 μL
4950
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.