Learn More
Occludin Rabbit, anti-Human, CoraLite Plus 488, Clone: 8B2, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech CL488-80545-20UL
Description
The Occludin antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Occludin. This antibody recognizes human, mouse, pig antigen. The Occludin antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| Occludin | |
| 8B2 | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Pig | |
| 150 kDa | |
| IgG |
| Immunofluorescence | |
| 1000 μg/mL | |
| occludin, OCLN | |
| 20 μL | |
| 4950 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |