Learn More
Occludin Rabbit, anti-Human, CoraLite Plus 488, Clone: 8B2, Proteintech
Rabbit Recombinant Monoclonal Antibody
€ 145.00 - € 450.00
Specifications
| Antigen | Occludin |
|---|---|
| Clone | 8B2 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18077275
|
Proteintech
CL488-80545-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18017815
|
Proteintech
CL488-80545-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Occludin antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Occludin. This antibody recognizes human, mouse, pig antigen. The Occludin antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| Occludin | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 4950 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 8B2 | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Pig | |
| occludin, OCLN | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.