missing translation for 'onlineSavingsMsg'
Learn More

Octhilinone, TRC

Product Code. 30005768
Change view
Click to view available options
Quantity:
250 mg
2.5 g
5 g
Unit Size:
2.5g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005768 5 g 5g
30006217 2.5 g 2.5g
18090040 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005768 Supplier Toronto Research Chemicals Supplier No. TRCO2393005G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26530-20-1
Molecular Formula C11 H19 N O S
Molecular Weight (g/mol) 213.34
Synonym 2-Octyl-2H-isothiazol-3-one, OIT
IUPAC Name 2-octyl-1,2-thiazol-3-one
SMILES CCCCCCCCN1SC=CC1=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C11H19NOS/c1-2-3-4-5-6-7-9-12-11(13)8-10-14-12/h8,10H,2-7,9H2,1H3
Formula Weight 213.1187
Percent Purity >95%
Chemical Name or Material Octhilinone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.