Learn More
Thermo Scientific™ Olympus™ 1.25X Objective, PlanApo N, 0.04 NA
Product Code. 17236583 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 each
missing translation for 'unitSize'
Each
Product Code. 17236583
missing translation for 'mfr': Thermo Scientific™ N010125
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Expand the functionality of your Thermo Scientific™ high-content instrument
Add a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific™ ArrayScan™ HCA Readers and the Thermo Scientific™ CellInsight™ NXT High Content Screening (HCS) Platform.
TRUSTED_SUSTAINABILITY
Specifications
Type Objective Lens
Product Line Olympus
Description Olympus™ 1.25X Objective, PlanApo N (0.04 NA)
For Use With (Equipment) ArrayScan HCA Readers
Lens Type Dry
Magnification 1.25x (0.04NA)
Quantity 1 each