Learn More
Thermo Scientific™ Open Top Screw Caps for Sample Storage Vials
Click to view available options
Closure Size:
8 mm
13 mm
15 mm
18 mm
20 mm
24 mm
Unit Size:
Pack of 100
Description
Open Top Screw Caps for Sample Storage Vials
Specifications
Specifications
| Closure Color | Black |
| Description | Open top 20-400 Screw Cap |
| Closure Material | Polypropylene |
| Closure Type | Screw Cap (Open-Top) |
| Quantity | 100 |
| Closure Size | 20 mm |