missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORAI3 Antibody (S06-8B3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19901-50ul
This item is not returnable.
View return policy
Description
ORAI3 Monoclonal antibody specifically detects ORAI3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ORAI3 | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry 1:100, Immunohistochemistry-Paraffin | |
| ORAI calcium release-activated calcium modulator 3, TMEM142C | |
| Recombinant protein of human Orai3 (Uniprot # Q9BRQ5) | |
| 50 μg | |
| Immunology | |
| 93129 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| S06-8B3 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction